Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Got2
Entrez Gene ID
33373
Gene Name
Glutamate oxaloacetate transaminase 2
Gene Aliases
152801_at, CG4233, Dmel\CG4233, Dmel_CG4233, Got, Got 2, Got-2, got2
Chromosome Location
Chr.2L: 1984512 - 1987758 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0001125
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_164445.4 NP_722744.1 3 - 4 432 77
NM_164446.1 NP_722745.1 3 - 4 417 77
GenBank mRNA BT125856.1 - 3 - 4 355 77

View Full Product Details