Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Got2
- Entrez Gene ID
-
33373
- Gene Name
- Glutamate oxaloacetate transaminase 2
- Gene Aliases
-
152801_at, CG4233, Dmel\CG4233, Dmel_CG4233, Got, Got 2, Got-2, got2
- Chromosome Location
-
Chr.2L: 1984512 - 1987758 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0001125
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_164446.1 |
NP_722745.1 |
1 - 2 |
151 |
58 |
GenBank mRNA |
BT125856.1 |
- |
1 - 2 |
89 |
58 |
View Full Product Details