Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Sodh-2
- Entrez Gene ID
-
41313
- Gene Name
- Sorbitol dehydrogenase-2
- Gene Aliases
-
CG4649, DM_7299382, Dmel\CG4649, Dmel_CG4649, SODH-2, Sdh, Sdh-2
- Chromosome Location
-
Chr.3R: 10876384 - 10878257 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0022359
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079587.3 |
NP_524311.1 |
5 - 6 |
967 |
71 |
GenBank mRNA |
AY058731.1 |
- |
5 - 6 |
955 |
71 |
View Full Product Details