Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cdk2
Entrez Gene ID
42453
Gene Name
Cyclin-dependent kinase 2
Gene Aliases
CDC2C, CDC2c, CDK2, CDK2/CDC2c, CG10498, Cdc2c, Dcdc2c, DmCdc2c, DmCdk2, Dmcdc2c, Dmel\CG10498, Dmel_CG10498, S(Sev-CycE)3A, cdc2c, cdk2, dCdk2
Chromosome Location
Chr.3R: 20735623 - 20738414 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0004107
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001170195.3 NP_001163666.1 2 - 3 394 133
NM_079696.6 NP_524420.1 2 - 3 394 133
NM_169916.3 NP_732544.1 2 - 3 394 133
GenBank mRNA AY051671.1 - 2 - 3 353 133
X57486.1 - - 397 133

View Full Product Details