Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rpn5
Entrez Gene ID
40717
Gene Name
Regulatory particle non-ATPase 5
Gene Aliases
CG1100, Dmel\CG1100, Dmel_CG1100, RPN5, p55, rpn5
Chromosome Location
Chr.3R: 5789493 - 5791569 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0028690
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_141331.3 NP_649588.1 2 - 3 1268 75
GenBank mRNA AF145315.1 - 1 - 2 1044 75
AY058339.1 - 2 - 3 1217 75

View Full Product Details