The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001202338.2 | NP_001189267.1 | 3 - 4 | 1812 | 105 |
NM_142822.3 | NP_651079.2 | 3 - 4 | 1815 | 105 |