Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- rha
- Entrez Gene ID
-
43071
- Gene Name
- CG11908-PA
- Gene Aliases
-
CG11908, Dmel\CG11908, Dmel_CG11908, RADHA, RHA, anon-WO0140519.50
- Chromosome Location
-
Chr.3R: 25340252 - 25344838 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0027376
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_143133.3 |
NP_651390.2 |
5 - 6 |
3996 |
81 |
View Full Product Details