Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lnk
Entrez Gene ID
43130
Gene Name
CG17367-PA
Gene Aliases
CG 17367, CG17367, CG5908, Dmel\CG17367, Dmel_CG17367, LNK, SH2B, dSH2B, lnk
Chromosome Location
Chr.3R: 25888542 - 25893270 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0028717
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001300607.1 NP_001287536.1 3 - 4 2311 93
NM_143181.3 NP_651438.1 3 - 4 2101 93
NM_170246.2 NP_733125.1 3 - 4 2296 93
NM_170247.2 NP_733126.1 3 - 4 2470 93
GenBank mRNA AY069404.1 - 3 - 4 2098 93

View Full Product Details