Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dgt1
Entrez Gene ID
43529
Gene Name
dim gamma-tubulin 1
Gene Aliases
CG18041, Dgt1, Dmel\CG18041, Dmel_CG18041, NSL2
Chromosome Location
Chr.3R: 29854695 - 29857491 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0039710
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001276154.1 NP_001263083.1 2 - 3 804 65
NM_001300666.1 NP_001287595.1 2 - 3 804 65
NM_143480.4 NP_651737.1 2 - 3 804 65
GenBank mRNA AY061156.1 - 2 - 3 784 65

View Full Product Details