Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gel
Entrez Gene ID
46008
Gene Name
Gelsolin
Gene Aliases
CG1106, DGS, Dmel\CG1106, Dmel_CG1106, gel
Chromosome Location
Chr.3R: 4235217 - 4241057 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0010225
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001043192.3 NP_001036657.2 2 - 3 304 83
NM_080126.4 NP_524865.2 3 - 4 556 83
NM_164319.4 NP_730788.1 2 - 3 348 83
NM_164321.5 NP_730790.2 4 - 5 750 83
NM_176394.3 NP_788571.1 3 - 4 527 83
NM_206426.4 NP_996148.2 3 - 4 559 83
NM_206427.4 NP_996149.3 4 - 5 617 83

View Full Product Details