Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cdep
Entrez Gene ID
40608
Gene Name
Chondrocyte-derived ezrin-like domain containing protein
Gene Aliases
BcDNA:AT26639, BcDNA:RH50880, CDEP, CG1283, CG2008, CG31536, CG34306, CG44193, Dmel\CG44193, Dmel_CG44193, cdep, i23
Chromosome Location
Chr.3R: 4910552 - 4945576 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0265082
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001043211.2 NP_001036676.1 12 - 13 4442 66
NM_169022.2 NP_730882.2 12 - 13 4430 66
GenBank mRNA BT024186.1 - 11 - 12 3432 66

View Full Product Details