Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cu
Entrez Gene ID
41339
Gene Name
curled
Gene Aliases
CG31299, CG4782, CG4796, Dmel\CG31299, Dmel_CG31299, no, no(cu), noc, noct, nocturnin
Chromosome Location
Chr.3R: 11197592 - 11208633 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0261808
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001104276.2 NP_001097746.1 1 - 2 753 77

View Full Product Details