Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Octbeta3R
Entrez Gene ID
3885573
Gene Name
Octopamine beta3 receptor
Gene Aliases
CG18554, CG31348, CG31350, CG31351, CG33959, CG34055, CG42244, CG7078, DmCG7078, DmOctbeta3R, Dmel\CG42244, Dmel_CG42244, OctBeta3, Octb3R, anon-WO0170980.187, anon-WO0170980.188, octRbeta3, octbeta3R
Chromosome Location
Chr.3R: 12511570 - 12548229 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0250910
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038954.2 NP_001034043.2 5 - 6 2078 65
NM_001038957.3 NP_001034046.3 5 - 6 2078 65
NM_001038959.3 NP_001034048.2 5 - 6 2078 65
NM_141953.3 NP_650210.2 5 - 6 2078 65

View Full Product Details