Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
beat-Vb
Entrez Gene ID
41583
Gene Name
beaten path Vb
Gene Aliases
Beat-Vb, CG14385, CG14386, CG31298, Dmel\CG31298, Dmel_CG31298, beat Vb
Chromosome Location
Chr.3R: 12916056 - 12934813 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0038092
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001275582.1 NP_001262511.1 6 - 7 1215 98
NM_169481.1 NP_731748.1 6 - 7 1410 98
GenBank mRNA AY070836.1 - - 1410 98

View Full Product Details