Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Atx2
- Entrez Gene ID
-
41883
- Gene Name
- Ataxin-2
- Gene Aliases
-
ATX2, Atx-2, Atxain-2, BEST:GH27029, CG43368, CG5166, CG5166a, DAtx2, Datx2, Dmel\CG5166, Dmel_CG5166, GH27029, SCA2, Sca2, anon-EST:fe2E1, anon-WO0118547.351, anon-WO0140519.1, atx-2, atx2, dAtx2, dSCA2, l(3)06490
- Chromosome Location
-
Chr.3R: 15408423 - 15417277 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0041188
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_169657.3 |
NP_732033.1 |
1 - 2 |
772 |
98 |
GenBank mRNA |
AY095516.1 |
- |
1 - 2 |
524 |
98 |
View Full Product Details