Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- bnl
- Entrez Gene ID
-
42356
- Gene Name
- branchless
- Gene Aliases
-
BNL, Bnl, Branchless, CG4608, DFGF, DmBnl, Dmel\CG4608, Dmel_CG4608, FGF, dFGF, l(3)06916, l(3)neo52
- Chromosome Location
-
Chr.3R: 19794073 - 19836817 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0014135
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_169874.1 |
NP_732452.1 |
4 - 5 |
3369 |
61 |
View Full Product Details