Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KrT95D
Entrez Gene ID
42843
Gene Name
Krueppel target at 95D
Gene Aliases
CG5405, Dmel\CG5405, Dmel_CG5405
Chromosome Location
Chr.3R: 23997538 - 24031827 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0020647
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001170238.2 NP_001163709.2 7 - 8 3660 99
NM_079749.6 NP_524473.2 8 - 9 3786 99
NM_170108.4 NP_732933.1 9 - 10 3106 99
NM_170109.2 NP_732934.2 6 - 7 2122 99
NM_170110.4 NP_732935.2 8 - 9 2980 99
GenBank mRNA AY061587.1 - 7 - 8 2010 99
AY119444.1 - 8 - 9 2963 99

View Full Product Details