Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pli
Entrez Gene ID
42821
Gene Name
Pellino
Gene Aliases
CG5212, Dmel\CG5212, Dmel_CG5212, PAP, PELLINO, pli
Chromosome Location
Chr.3R: 23860162 - 23890052 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0025574
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001104420.3 NP_001097890.1 3 - 4 511 62
NM_079742.4 NP_524466.1 3 - 4 583 62
GenBank mRNA AF091624.1 - 3 - 4 488 62
AY069632.1 - 3 - 4 559 62

View Full Product Details