Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pp1alpha-96A
Entrez Gene ID
42922
Gene Name
Protein phosphatase 1alpha at 96A
Gene Aliases
96A, CG6593, DmPp1-96A, Dmel\CG6593, Dmel_CG6593, PP1, PP1 96A, PP1 alpha96a, PP1A-96A, PP1alpha, PP1alpha-96A, PP1alpha96A, PP1alpha[[1]], PP1c, PP1c 96A, Pp1-96A, pp1alpha-96A, pp1alpha96A
Chromosome Location
Chr.3R: 24518270 - 24522331 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0003134
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001275990.1 NP_001262919.1 2 - 3 782 67
NM_079760.3 NP_524484.1 2 - 3 782 67
GenBank mRNA BT016110.1 - 2 - 3 762 67
X56438.1 - - 778 67

View Full Product Details