Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cyp4c3
Entrez Gene ID
43663
Gene Name
Cytochrome P450-4c3
Gene Aliases
4C3, CG1438, Dmel\CG1438, Dmel_CG1438, P-450, P450, cyp4c3
Chromosome Location
Chr.3R: 30894038 - 30900688 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0015032
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_079859.4 NP_524598.1 2 - 3 482 68
GenBank mRNA BT010108.2 - 2 - 3 559 68

View Full Product Details