Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Gr98b
- Entrez Gene ID
-
117327
- Gene Name
- Gustatory receptor 98b
- Gene Aliases
-
98B.2, CG12884, CG31059, Dmel\CG31059, Dmel_CG31059, GR98B.2, GR98b, Gr98A1, Gr98B2
- Chromosome Location
-
Chr.3R: 27542497 - 27543888 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0046887
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_170334.1 |
NP_733213.1 |
2 - 3 |
953 |
91 |
View Full Product Details