Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cep43
Entrez Gene ID
415215
Gene Name
centrosomal protein 43
Gene Aliases
ccr6a, fgfr1op, zgc:86782
Chromosome Location
Chr.20: 30578966 - 30610172 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002125.1 NP_001002125.1 3 - 4 213 116
XM_005160662.4 XP_005160719.1 3 - 4 247 116
XM_005160663.4 XP_005160720.1 3 - 4 247 116
XM_005160664.4 XP_005160721.1 3 - 4 247 116
XM_017352461.2 XP_017207950.1 3 - 4 247 116
XM_021468390.1 XP_021324065.1 3 - 4 247 116
XM_021468391.1 XP_021324066.1 3 - 4 247 116
XM_021468392.1 XP_021324067.1 3 - 4 247 116
XM_021468393.1 XP_021324068.1 3 - 4 247 116
GenBank mRNA BC071450.1 - 3 - 4 213 116

View Full Product Details