Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- mzt2b
- Entrez Gene ID
-
445174
- Gene Name
- mitotic spindle organizing protein 2B
- Gene Aliases
-
fam128, mzt2, zgc:101080
- Chromosome Location
-
Chr.21: 16079123 - 16087880 on Build GRCz11
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC078275.1 |
- |
2 - 3 |
249 |
101 |
View Full Product Details