Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
vtna
Entrez Gene ID
553699
Gene Name
vitronectin a
Gene Aliases
zgc:112277
Chromosome Location
Chr.15: 28199341 - 28208938 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001020672.1 NP_001018508.1 5 - 6 837 84
XM_021481411.1 XP_021337086.1 6 - 7 948 84
GenBank mRNA BC095724.1 - 5 - 6 837 84

View Full Product Details