Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rhot1b
Entrez Gene ID
561933
Gene Name
ras homolog family member T1
Gene Aliases
miro1, rhot1, si:dkeyp-97g3.6
Chromosome Location
Chr.6: 100535 - 126075 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082826.2 NP_001076295.2 12 - 13 1181 140
XM_005166191.4 XP_005166248.1 12 - 13 1206 140
XM_021476893.1 XP_021332568.1 11 - 12 1107 140
XM_021476894.1 XP_021332569.1 12 - 13 1209 140
XM_021476895.1 XP_021332570.1 6 - 7 538 140
GenBank mRNA BC163039.1 - 12 - 13 1181 140

View Full Product Details