Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ubr7
Entrez Gene ID
322815
Gene Name
ubiquitin protein ligase E3 component n-recognin 7
Gene Aliases
fb74a06, wu:fb74a06, wu:fi29b06, zgc:55404
Chromosome Location
Chr.20: 27092665 - 27107703 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_212629.1 NP_997794.1 5 - 6 677 82
XM_021468365.1 XP_021324040.1 5 - 6 1089 82
GenBank mRNA BC044390.1 - 5 - 6 677 82

View Full Product Details