The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001001889.1 | NP_001001889.1 | 14 - 15 | 2300 | 57 |
GenBank mRNA | AF329680.1 | - | 14 - 15 | 2300 | 57 |
BC056276.1 | - | 14 - 15 | 2297 | 57 | |
BC114257.1 | - | 7 - 8 | 1049 | 57 | |
BC134896.1 | - | 6 - 7 | 651 | 57 |