Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
park7
Entrez Gene ID
449674
Gene Name
parkinson protein 7
Gene Aliases
dj1, zgc:103725
Chromosome Location
Chr.11: 41459837 - 41474162 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005938.1 NP_001005938.1 2 - 3 252 80
GenBank mRNA BC083475.1 - 2 - 3 252 80
DQ882651.1 - 2 - 3 259 80

View Full Product Details