Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tomm40
Entrez Gene ID
322645
Gene Name
translocase of outer mitochondrial membrane 40 homolog (yeast)
Gene Aliases
wu:fb68h07, zgc:64168
Chromosome Location
Chr.16: 25672639 - 25680698 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199614.1 NP_955908.1 5 - 6 510 70
XM_005158091.4 XP_005158148.1 5 - 6 918 70
XM_017351318.2 XP_017206807.1 5 - 6 707 70
GenBank mRNA BC053295.1 - 5 - 6 510 70

View Full Product Details