Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
skor1b
Entrez Gene ID
567457
Gene Name
SKI family transcriptional corepressor 1b
Gene Aliases
lbxcor1, si:dkey-246a16.3, skor1
Chromosome Location
Chr.18: 168179 - 173867 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045133.1 NP_001038598.1 4 - 5 2004 93
XM_005159134.4 XP_005159191.1 5 - 6 2529 93
XM_005159135.4 XP_005159192.1 5 - 6 2528 93
XM_005159136.4 XP_005159193.1 5 - 6 2522 93
GenBank mRNA BC134846.1 - 5 - 6 2306 93

View Full Product Details