Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ache
Entrez Gene ID
114549
Gene Name
acetylcholinesterase
Gene Aliases
zgc:92550
Chromosome Location
Chr.7: 26077153 - 26087817 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131846.2 NP_571921.1 2 - 3 1677 68
GenBank mRNA BC163891.1 - 2 - 3 1677 68
BC163898.1 - 2 - 3 1677 68

View Full Product Details