Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
aqp4
Entrez Gene ID
445293
Gene Name
aquaporin 4
Gene Aliases
si:ch211-192k9.1, zgc:100922
Chromosome Location
Chr.20: 18163368 - 18189965 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003749.4 NP_001003749.1 1 - 2 67 71
NM_001358313.1 NP_001345242.1 1 - 2 67 71
GenBank mRNA BC078213.1 - 1 - 2 54 71
CK682060.1 - 1 - 2 85 71

View Full Product Details