Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
evx1
Entrez Gene ID
30499
Gene Name
even-skipped homeobox 1
Gene Aliases
cb416, evx-1
Chromosome Location
Chr.19: 19717971 - 19721556 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131249.2 NP_571324.2 2 - 3 764 65
GenBank mRNA AF305889.1 - 1 - 2 807 65
BC078301.1 - 2 - 3 360 65

View Full Product Details