Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ncam1b
Entrez Gene ID
114442
Gene Name
neural cell adhesion molecule 1b
Gene Aliases
ncam3, zPCAM
Chromosome Location
Chr.15: 18367307 - 18574863 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131831.1 NP_571906.1 16 - 17 2324 107
XM_005157405.4 XP_005157462.1 20 - 21 2543 107
XM_005157406.4 XP_005157463.1 19 - 20 2519 107
XM_005157407.4 XP_005157464.1 18 - 19 2495 107
XM_005157408.4 XP_005157465.1 17 - 18 2492 107
XM_005157409.4 XP_005157466.1 18 - 19 2489 107
XM_005157410.4 XP_005157467.1 16 - 17 2467 107
XM_005157411.4 XP_005157468.1 15 - 16 2437 107
XM_009291549.3 XP_009289824.1 19 - 20 2540 107
XM_009291550.3 XP_009289825.1 19 - 20 2528 107
XM_009291551.3 XP_009289826.1 18 - 19 2525 107
XM_009291552.3 XP_009289827.1 19 - 20 2513 107
XM_009291553.3 XP_009289828.1 19 - 20 2510 107
XM_009291554.3 XP_009289829.1 18 - 19 2507 107
XM_009291556.3 XP_009289831.1 20 - 21 2543 107
XM_009291557.3 XP_009289832.1 20 - 21 2543 107
XM_009291559.3 XP_009289834.1 16 - 17 2441 107
XM_017359195.2 XP_017214684.1 18 - 19 2504 107
GenBank mRNA AY029401.1 - 16 - 17 2324 107
BC162073.1 - 16 - 17 2224 107
CU458747.1 - 16 - 17 2111 107
HM467819.1 - 16 - 17 2114 107

View Full Product Details