Assay Design
The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.
Gene Transcripts
- Gene Symbol
- nell2a
- Entrez Gene ID
-
790930
- Gene Name
- neural EGFL like 2a
- Gene Aliases
-
nell2, wu:fj43h11, zgc:158375
- Chromosome Location
-
Chr.25: 581140 - 692366 on Build GRCz11
- Gene Symbol
- nell2b
- Entrez Gene ID
-
336082
- Gene Name
- neural EGFL like 2b
- Gene Aliases
-
fj47f01, si:dkey-234l24.6, wu:fj47f01
- Chromosome Location
-
Chr.4: 14247745 - 14328978 on Build GRCz11
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
CA476850.1 |
- |
- |
239 |
70 |
View Full Product Details