Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
sept5a
Entrez Gene ID
335925
Gene Name
septin 5a
Gene Aliases
fj37h04, sept5, wu:fj37h04, zgc:73218
Chromosome Location
Chr.8: 4588886 - 4618586 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199988.1 NP_956282.1 8 - 9 765 137
GenBank mRNA BC059564.1 - 8 - 9 765 137

View Full Product Details