Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
foxp3a
Entrez Gene ID
571165
Gene Name
forkhead box P3a
Gene Aliases
foxp3
Chromosome Location
Chr.8: 25716307 - 25731177 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001329567.1 NP_001316496.1 11 - 12 1086 100
XM_021478298.1 XP_021333973.1 11 - 12 1142 100
GenBank mRNA BC154484.1 - 11 - 12 1086 100
BC163356.1 - 10 - 11 1065 100
FJ906821.1 - 10 - 11 992 100
FM881778.1 - 10 - 11 1059 100
FM881779.1 - 6 - 7 636 100

View Full Product Details