Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
ppid
Entrez Gene ID
415155
Gene Name
peptidylprolyl isomerase D
Gene Aliases
wu:fb18b07, zgc:86711
Chromosome Location
Chr.14: 48045193 - 48065365 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002065.1 NP_001002065.1 5 - 6 741 111
GenBank mRNA BC071388.1 - 5 - 6 741 111

View Full Product Details