Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
sgk2b
Entrez Gene ID
559050
Gene Name
serum/glucocorticoid regulated kinase 2b
Gene Aliases
zgc:154065
Chromosome Location
Chr.16: 26530426 - 26537126 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076564.1 NP_001070032.1 2 - 3 166 76
GenBank mRNA BC124529.1 - 2 - 3 166 76
BC164500.1 - 2 - 3 150 76

View Full Product Details