Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tbx5a
Entrez Gene ID
30071
Gene Name
T-box transcription factor 5a
Gene Aliases
hst, tbx5, tbx5.1
Chromosome Location
Chr.5: 72264163 - 72303894 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF152607.1 - 3 - 4 405 102
AF179407.1 - 3 - 4 648 102
AF185283.1 - 2 - 3 150 102

View Full Product Details