Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tfr1b
Entrez Gene ID
494478
Gene Name
transferrin receptor 1b
Gene Aliases
-
Chromosome Location
Chr.24: 26017094 - 26031250 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009918.2 NP_001009918.2 16 - 17 2072 101
GenBank mRNA AY649364.1 - 15 - 16 1912 101
BC108025.1 - 16 - 17 2072 101

View Full Product Details