Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
msrb3
Entrez Gene ID
415184
Gene Name
methionine sulfoxide reductase B3
Gene Aliases
wu:fc07g04, zgc:86909
Chromosome Location
Chr.4: 12892466 - 12914192 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002094.2 NP_001002094.2 6 - 7 602 82
NM_001320378.1 NP_001307307.1 6 - 7 635 82
NM_001320380.1 NP_001307309.1 5 - 6 448 82
XM_009300117.2 XP_009298392.1 6 - 7 559 82
XM_021474986.1 XP_021330661.1 7 - 8 767 82
GenBank mRNA BC071530.1 - 6 - 7 559 82

View Full Product Details