Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
olfm1a
Entrez Gene ID
436764
Gene Name
olfactomedin 1a
Gene Aliases
zgc:111962, zgc:92854
Chromosome Location
Chr.5: 64731808 - 64779969 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002491.2 NP_001002491.1 1 - 2 305 80
XM_005165598.4 XP_005165655.1 1 - 2 533 80
GenBank mRNA BC076316.1 - 1 - 2 309 80
EH445468.1 - 1 - 2 331 80
EH471345.1 - 3 - 4 615 80

View Full Product Details