Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
osbpl3a
Entrez Gene ID
447908
Gene Name
oxysterol binding protein-like 3a
Gene Aliases
osbpl3, zgc:101089
Chromosome Location
Chr.19: 19989343 - 20032761 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004646.1 NP_001004646.1 9 - 10 894 146
XM_005170028.4 XP_005170085.1 9 - 10 1148 146
GenBank mRNA BC078429.1 - 9 - 10 894 146

View Full Product Details