Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
st6galnac5b
Entrez Gene ID
407716
Gene Name
ST6 (alpha-N-acetyl-neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminide alpha-2,6-sialyltransferase 5b
Gene Aliases
siat7E, st6galnac5, st6galnac5-r1, zgc:92262
Chromosome Location
Chr.8: 319910 - 343329 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006662.2 NP_001006663.2 2 - 3 340 123
XM_021478236.1 XP_021333911.1 2 - 3 287 123
GenBank mRNA AJ646874.1 - 1 - 2 198 123
BC081635.1 - 2 - 3 317 123

View Full Product Details