Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
blvra
Entrez Gene ID
767661
Gene Name
biliverdin reductase A
Gene Aliases
zgc:153046
Chromosome Location
Chr.2: 49844630 - 49860704 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076601.1 NP_001070069.1 2 - 3 232 115
GenBank mRNA BC124225.1 - 2 - 3 232 115
BC163949.1 - 1 - 2 150 115

View Full Product Details