Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rhbg
Entrez Gene ID
337596
Gene Name
Rh family B glycoprotein
Gene Aliases
fj85f07, wu:fj85f07, zgc:56354
Chromosome Location
Chr.16: 29265801 - 29277186 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200071.3 NP_956365.2 4 - 5 746 69
GenBank mRNA AF529360.1 - 4 - 5 681 69
BC049405.1 - 4 - 5 698 69
EH442413.1 - 4 - 5 772 69

View Full Product Details