Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nrxn1a
Entrez Gene ID
565531
Gene Name
neurexin 1a
Gene Aliases
-
Chromosome Location
Chr.12: 114926 - 403053 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080021.2 NP_001073490.1 11 - 12 3297 83
XM_005156209.4 XP_005156266.1 11 - 12 3279 83
XM_009306641.3 XP_009304916.1 11 - 12 3279 83
XM_009306642.3 XP_009304917.1 11 - 12 3276 83
XM_009306643.3 XP_009304918.1 11 - 12 3278 83
XM_009306644.3 XP_009304919.1 11 - 12 3279 83
XM_009306645.3 XP_009304920.1 10 - 11 3267 83
XM_009306646.3 XP_009304921.1 10 - 11 3264 83
XM_009306648.3 XP_009304923.1 10 - 11 3261 83
XM_009306649.3 XP_009304924.1 10 - 11 3255 83
XM_009306651.3 XP_009304926.1 11 - 12 3278 83
XM_009306652.3 XP_009304927.1 11 - 12 3279 83
XM_009306653.3 XP_009304928.1 11 - 12 3279 83
XM_009306654.3 XP_009304929.1 11 - 12 3279 83
XM_009306656.3 XP_009304931.1 11 - 12 3279 83
XM_021480136.1 XP_021335811.1 9 - 10 3249 83
XM_021480137.1 XP_021335812.1 11 - 12 3278 83
XM_021480138.1 XP_021335813.1 8 - 9 3219 83
XM_021480140.1 XP_021335815.1 9 - 10 3242 83
XM_021480143.1 XP_021335818.1 9 - 10 3248 83
GenBank mRNA DQ641424.1 - 10 - 11 3246 83

View Full Product Details