Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ap4b1
Entrez Gene ID
393309
Gene Name
adaptor related protein complex 4 subunit beta 1
Gene Aliases
ap4b1l, zgc:63878
Chromosome Location
Chr.23: 354369 - 367693 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200338.1 NP_956632.1 2 - 3 207 93
XM_009296550.3 XP_009294825.1 2 - 3 256 93
GenBank mRNA BC053128.1 - 2 - 3 207 93

View Full Product Details