Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
sav1
Entrez Gene ID
447821
Gene Name
salvador family WW domain containing protein 1
Gene Aliases
zgc:92250
Chromosome Location
Chr.13: 36787077 - 36798204 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004560.1 NP_001004560.1 2 - 3 882 83
GenBank mRNA BC081632.1 - 2 - 3 882 83

View Full Product Details